Gene Transcripts

Gene Symbol
SLC10A5
Entrez Gene ID
347051
Gene Name
solute carrier family 10 member 5
Gene Aliases
P5
Chromosome Location
Chr. 8: 81693656 - 81694972 on Build GRCh38
UniGene ID
Hs.683849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010893.2 NP_001010893.1 1 67
GenBank mRNA AK095808.1 - 1 1272
AY825924.1 - 1 67
BC136625.1 - 1 147 9020615
BC136631.1 - 1 147 9020621

View Full Product Details