Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000447.2 | NP_000438.2 | 11 | 1410 | |
NM_012486.2 | NP_036618.2 | 10, 11 | 1407 | ||
XM_005273199.3 | XP_005273256.1 | 10 | 1223 | ||
XM_017001835.1 | XP_016857324.1 | 10 | 1731 | ||
XM_017001836.1 | XP_016857325.1 | 9, 10 | 1220 | ||
GenBank mRNA | AF416718.1 | - | 9 | 1051 | |
AK292299.1 | - | 11, 12 | 1359 | ||
AK301143.1 | - | 10 | 1159 | ||
BC006365.2 | - | 12 | 1604 | 4131408 | |
BT006984.1 | - | 9 | 983 | ||
L43964.1 | - | 10 | 1350 | ||
L44577.1 | - | 11 | 1348 | ||
U34349.1 | - | 8, 9 | 1071 |