Gene Transcripts
- Gene Symbol
- ITGB3
- Entrez Gene ID
- 3690
- Gene Name
- integrin subunit beta 3
- Gene Aliases
- BDPLT16, BDPLT2, CD61, GP3A, GPIIIa, GT
- Chromosome Location
- Chr. 17: 47253842 - 47312711 on Build GRCh38
- UniGene ID
- Hs.218040
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000212.2 |
NP_000203.2
|
8, 9 |
1138 |
|
GenBank mRNA |
AY826979.1 |
- |
8, 9 |
1127 |
|
BC127666.1 |
- |
8, 9 |
1138 |
40128462 |
BC127667.1 |
- |
8, 9 |
1142 |
40128464 |
J02703.1 |
- |
8, 9 |
1138 |
|
KC120777.1 |
- |
8, 9 |
1138 |
|
KC120778.1 |
- |
8, 9 |
1138 |
|
M20311.1 |
- |
8, 9 |
1134 |
|
M35999.1 |
- |
8, 9 |
1136 |
|
U95204.1 |
- |
8, 9 |
1118 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU174383.1 |
- |
1, 2 |
90 |
|
Chr. 17 - 47290183 - 47291488 [+] |
View Full Product Details