Gene Transcripts

Gene Symbol
SYCE3
Entrez Gene ID
644186
Gene Name
synaptonemal complex central element protein 3
Gene Aliases
C22orf41, THEG2
Chromosome Location
Chr. 22: 50551112 - 50562899 on Build GRCh38
UniGene ID
Hs.535660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123225.1 NP_001116697.1 2 103
GenBank mRNA BC127859.1 - 1 88
BC127860.1 - 1 88 40134499
EU082028.1 - 1 61

View Full Product Details