Gene Transcripts

Gene Symbol
C15orf56
Entrez Gene ID
644809
Gene Name
chromosome 15 open reading frame 56
Gene Aliases
Chromosome Location
Chr. 15: 40250664 - 40252909 on Build GRCh38
UniGene ID
Hs.631718
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039905.2 NP_001034994.1 2 1266
GenBank mRNA AK095915.1 - 2 1266

View Full Product Details