Gene Transcripts

Gene Symbol
Hnrnpl
Entrez Gene ID
15388
Gene Name
heterogeneous nuclear ribonucleoprotein L
Gene Aliases
C79783, D830027H13Rik, Hnrpl
Chromosome Location
Chr. 7: 28808546 - 28822268 on Build GRCm38
UniGene ID
Mm.9043
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177301.5 NP_796275.3 3, 4 643
GenBank mRNA AK044748.1 - 3, 4 419
AK085906.1 - 3, 4 594
AK144684.1 - 3, 4 597
BC027206.1 - 3, 4 620 4218471
BC099683.1 - 3, 4 638 6530863

View Full Product Details