Gene Transcripts

Gene Symbol
Ufd1l
Entrez Gene ID
22230
Gene Name
ubiquitin fusion degradation 1 like
Gene Aliases
Ufd1
Chromosome Location
Chr. 16: 18811801 - 18835261 on Build GRCm38
UniGene ID
Mm.237594
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011672.4 NP_035802.3 10 824
GenBank mRNA AK010461.1 - 10 793
AK012543.1 - 8 768
AK088823.1 - 10 793
AK147138.1 - 10 807
AK169062.1 - 10 806
BC006630.1 - 10 781 3708320
U64445.1 - 10 809

View Full Product Details