Gene Transcripts

Gene Symbol
Jph1
Entrez Gene ID
57339
Gene Name
junctophilin 1
Gene Aliases
ENSMUSG00000054314, JP-1, Jp1
Chromosome Location
Chr. 1: 16942853 - 17097932 on Build GRCm38
UniGene ID
Mm.143761
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020604.2 NP_065629.1 4 1770
GenBank mRNA AB024445.1 - 4 1679
BC120839.1 - 4 1770 40129762
BC137669.1 - 4 1770 8860688

View Full Product Details