Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289514.1 | NP_001276443.1 | 15 | 2357 | |
NM_019636.3 | NP_062610.2 | 16 | 2938 | ||
GenBank mRNA | AK122445.1 | - | 18 | 4026 | |
AK135665.1 | - | 15 | 2529 | ||
AK148356.1 | - | 15 | 2314 | ||
AK154591.1 | - | 17 | 2974 | ||
AK164923.1 | - | 12 | 1642 | ||
AK168763.1 | - | 5 | 643 | ||
BC004675.1 | - | 5 | 596 | ||
BC059824.1 | - | 16 | 3112 | ||
BC117931.1 | - | 15 | 2606 | 40097712 | |
U33005.1 | - | 16 | 2894 |