Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025417.2 | NP_079693.1 | 6 | 348 | |
GenBank mRNA | AK004174.1 | - | 6 | 349 | |
AK009802.1 | - | 6 | 347 | ||
AK172194.1 | - | 5 | 1460 | ||
BC033438.1 | - | 5 | 334 | 3995599 | |
BC049222.1 | - | 6 | 347 | 2649359 | |
BC055798.1 | - | 5 | 1439 |