Gene Transcripts

Gene Symbol
Rasl11a
Entrez Gene ID
68895
Gene Name
RAS-like, family 11, member A
Gene Aliases
1110065D03Rik, AI852031
Chromosome Location
Chr. 5: 146845071 - 146847726 on Build GRCm38
UniGene ID
Mm.266978
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026864.1 NP_081140.1 4 580
GenBank mRNA AK004371.1 - 4 534
BC118009.1 - 1695 40102480

View Full Product Details