Gene Transcripts

Gene Symbol
Fra10ac1
Entrez Gene ID
70567
Gene Name
FRA10AC1 homolog (human)
Gene Aliases
5730455O13Rik, AI448293
Chromosome Location
Chr. 19: 38188477 - 38224513 on Build GRCm38
UniGene ID
Mm.30607
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081075.1 NP_001074544.1 4 373
GenBank mRNA AK017666.1 - 4 435
AK077572.1 - 4 346
AK172241.1 - 4 380

View Full Product Details