Gene Transcripts

Gene Symbol
Sval1
Entrez Gene ID
71578
Gene Name
seminal vesicle antigen-like 1
Gene Aliases
9130404C18Rik, SLP-C
Chromosome Location
Chr. 6: 41951628 - 41956098 on Build GRCm38
UniGene ID
Mm.96996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027832.3 NP_082108.1 3 297
GenBank mRNA AK018661.1 - 3 299
BC114422.1 - 3 292 40043764

View Full Product Details