Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009620.1 | NP_033750.1 | 1 | 1539 | |
GenBank mRNA | AK076647.1 | - | 1 | 1600 | |
AY158688.1 | - | 1 | 1539 | ||
BC150159.1 | - | 1 | 1543 | 40141514 | |
BC150481.1 | - | 1 | 1543 | 40141515 | |
U22058.1 | - | 1 | 636 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001038995.2 | NP_001034084.2 | 1 | 1598 | |
GenBank mRNA | BC141094.1 | - | 1 | 1576 | |
KF144384.1 | - | 1 | 249 | ||
KF144385.1 | - | 1 | 249 |