Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028500.3 | NP_082776.2 | 4, 5 | 595 | |
NM_029782.3 | NP_084058.2 | 2, 3 | 296 | ||
GenBank mRNA | AB206786.1 | - | 4, 5 | 500 | |
AK006582.1 | - | 4, 5 | 596 | ||
AK018263.1 | - | 2, 3 | 218 | ||
AK046081.1 | - | 2, 3 | 297 |