Gene Transcripts

Gene Symbol
Cep112
Entrez Gene ID
76380
Gene Name
centrosomal protein 112
Gene Aliases
1700001M19Rik, 1700029K01Rik, 8430407H02Rik, AV043680, AV207351, Ccdc46, Macoco
Chromosome Location
Chr. 11: 108425068 - 108860615 on Build GRCm38
UniGene ID
Mm.344071
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029586.2 NP_083862.1 2, 3 350
NM_029606.3 NP_083882.3 25, 26 2897
NM_145688.2 NP_663721.2 4, 5 572
GenBank mRNA AK005611.1 - 2, 3 351
AK006504.1 - 4, 5 558
BC005723.1 - 4, 5 384
BC048677.1 - 4, 5 544
BC086808.1 - 19, 20 2137 30632964
CA464637.1 - 4, 5 574

View Full Product Details