Gene Transcripts

Gene Symbol
Orm2
Entrez Gene ID
18406
Gene Name
orosomucoid 2
Gene Aliases
Agp1, Orm-2
Chromosome Location
Chr. 4: 63362449 - 63365877 on Build GRCm38
UniGene ID
Mm.14173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011016.2 NP_035146.1 1 110
GenBank mRNA BC057985.1 - 1 85
CB949119.1 - 1 113
M27009.1 - 1 75

View Full Product Details