Gene Transcripts

Gene Symbol
Ndufa3
Entrez Gene ID
66091
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3
Gene Aliases
1010001M12Rik, 1700022J01Rik
Chromosome Location
Chr. 7: 3617373 - 3620161 on Build GRCm38
UniGene ID
Mm.17851
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025348.2 NP_079624.1 4 263
GenBank mRNA AK003133.1 - 4 254
AK019099.1 - 4 265
BC061002.1 - 256
BC120712.1 - 4 257 40129635
BC120714.1 - 4 257 40129637

View Full Product Details