Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164741.1 | NP_001158213.1 | 8 | 1146 | |
NM_001666.4 | NP_001657.3 | 7 | 1026 | ||
GenBank mRNA | AK294562.1 | - | 7 | 951 | |
AY183123.1 | - | 7 | 1010 | ||
BC052303.1 | - | 8 | 1146 | 6379390 | |
D50921.1 | - | 7 | 957 | ||
X78817.1 | - | 7 | 1010 |