Gene Transcripts

Gene Symbol
ARSD
Entrez Gene ID
414
Gene Name
arylsulfatase D
Gene Aliases
ASD
Chromosome Location
Chr. X: 2903970 - 2929375 on Build GRCh38
UniGene ID
Hs.528631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001669.3 NP_001660.2 5 561
NM_009589.3 NP_033667.2 5 561
XM_005274514.2 XP_005274571.1 5 531
XM_005274515.2 XP_005274572.1 5 531
GenBank mRNA AF160499.1 - 5 461
AK314479.1 - 5 525
BC020229.1 - 489
X83572.1 - 5 537

View Full Product Details