Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033274.4 | NP_150377.1 | 6 | 603 | |
XM_005266003.2 | XP_005266060.1 | 6 | 555 | ||
GenBank mRNA | AF134707.1 | - | 3 | 184 | |
AF311317.1 | - | 6 | 512 | ||
AF326918.1 | - | 6 | 524 | ||
AK091464.1 | - | 6 | 555 | ||
AK308182.1 | - | 6 | 555 | ||
Y13786.2 | - | 6 | 553 |