Gene Transcripts

Gene Symbol
CAPN7
Entrez Gene ID
23473
Gene Name
calpain 7
Gene Aliases
CALPAIN7, PALBH
Chromosome Location
Chr. 3: 15206226 - 15252916 on Build GRCh38
UniGene ID
Hs.595234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014296.2 NP_055111.1 10 1431
XM_011533555.2 XP_011531857.1 11 1330
XM_011533556.2 XP_011531858.1 12 1562
XM_011533557.2 XP_011531859.1 11 1452
XM_017006043.1 XP_016861532.1 11 1514
XM_017006044.1 XP_016861533.1 10 1404
XM_017006045.1 XP_016861534.1 11 1470
GenBank mRNA AB028639.1 - 10 1412
AK314253.1 - 10 1393
BC056202.1 - 10 1409 5274889

View Full Product Details