Gene Transcripts

Gene Symbol
ANK2
Entrez Gene ID
287
Gene Name
ankyrin 2, neuronal
Gene Aliases
ANK-2, LQT4, brank-2
Chromosome Location
Chr. 4: 112706083 - 113383740 on Build GRCh38
UniGene ID
Hs.620557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127493.1 NP_001120965.1 19 2150
NM_001148.4 NP_001139.3 18 2092
NM_020977.3 NP_066187.2 18 2092
XM_005262945.2 XP_005263002.1 18 2190
XM_017008067.1 XP_016863556.1 18 2959
XM_017008068.1 XP_016863557.1 19 3200
XM_017008069.1 XP_016863558.1 17 2934
XM_017008070.1 XP_016863559.1 19 3199
XM_017008071.1 XP_016863560.1 19 2128
XM_017008072.1 XP_016863561.1 18 2104
XM_017008073.1 XP_016863562.1 18 2190
XM_017008074.1 XP_016863563.1 19 2128
XM_017008075.1 XP_016863564.1 19 2128
XM_017008076.1 XP_016863565.1 18 2190
XM_017008077.1 XP_016863566.1 18 2104
XM_017008078.1 XP_016863567.1 18 2190
XM_017008079.1 XP_016863568.1 17 2019
XM_017008080.1 XP_016863569.1 18 2104
XM_017008081.1 XP_016863570.1 18 2190
XM_017008082.1 XP_016863571.1 18 2334
XM_017008083.1 XP_016863572.1 18 2334
XM_017008084.1 XP_016863573.1 18 2190
XM_017008085.1 XP_016863574.1 18 2190
XM_017008086.1 XP_016863575.1 18 2190
XM_017008087.1 XP_016863576.1 18 2334
XM_017008088.1 XP_016863577.1 19 2128
XM_017008089.1 XP_016863578.1 18 2334
XM_017008090.1 XP_016863579.1 18 2334
XM_017008091.1 XP_016863580.1 17 2091
XM_017008092.1 XP_016863581.1 19 2128
XM_017008093.1 XP_016863582.1 19 2128
XM_017008094.1 XP_016863583.1 18 2190
XM_017008095.1 XP_016863584.1 18 2104
XM_017008096.1 XP_016863585.1 19 2128
XM_017008097.1 XP_016863586.1 18 2029
XM_017008098.1 XP_016863587.1 19 2128
XM_017008099.1 XP_016863588.1 19 2128
XM_017008100.1 XP_016863589.1 18 2029
XM_017008101.1 XP_016863590.1 18 2190
XM_017008102.1 XP_016863591.1 17 2005
XM_017008103.1 XP_016863592.1 18 2190
XM_017008104.1 XP_016863593.1 19 2128
XM_017008105.1 XP_016863594.1 18 2104
XM_017008106.1 XP_016863595.1 17 2091
XM_017008107.1 XP_016863596.1 18 2029
XM_017008108.1 XP_016863597.1 16 1992
XM_017008109.1 XP_016863598.1 18 2104
XM_017008110.1 XP_016863599.1 19 2128
XM_017008111.1 XP_016863600.1 18 2104
XM_017008112.1 XP_016863601.1 16 1906
GenBank mRNA AK095596.1 - 19 2150
AK299767.1 - 19 2116
AK299815.1 - 19 2128
AK307601.1 - 18 2043
BX537758.1 - 19 2124
X56958.2 - 18 2007
Z26634.2 - 18 1992

View Full Product Details