Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015305.3 | NP_056120.2 | 9 | 1888 | |
XM_017021115.1 | XP_016876604.1 | 10 | 1936 | ||
GenBank mRNA | AB018302.1 | - | 9 | 1785 | |
AK301589.1 | - | 5 | 455 | ||
AL137268.1 | - | 9 | 1786 | ||
BC028714.2 | - | 9 | 1835 | 4830897 |