Gene Transcripts
- Gene Symbol
- ACOXL
- Entrez Gene ID
- 55289
- Gene Name
- acyl-CoA oxidase-like
- Gene Aliases
- Chromosome Location
- Chr. 2: 110732539 - 111118571 on Build GRCh38
- UniGene ID
- Hs.253320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001904.1 |
- |
13 |
1603 |
|
View Full Product Details