Gene Transcripts

Gene Symbol
LANCL2
Entrez Gene ID
55915
Gene Name
LanC like 2
Gene Aliases
GPR69B, TASP
Chromosome Location
Chr. 7: 55365448 - 55433742 on Build GRCh38
UniGene ID
Hs.595384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018697.3 NP_061167.1 9 4097
GenBank mRNA AJ278245.1 - 9 3705
BC070049.1 - 9 3793 30343188

View Full Product Details