Gene Transcripts

Gene Symbol
BTBD1
Entrez Gene ID
53339
Gene Name
BTB domain containing 1
Gene Aliases
C15orf1, NS5ATP8
Chromosome Location
Chr. 15: 83016423 - 83067434 on Build GRCh38
UniGene ID
Hs.459149
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011885.1 NP_001011885.1 6, 7 1414
NM_025238.3 NP_079514.1 7, 8 1502
GenBank mRNA AF257241.1 - 7, 8 1381
AF355402.1 - 7, 8 1382
AF529369.1 - 7, 8 1299
AK000731.1 - 7, 8 1366
AK225355.1 - 7, 8 1366
AK225362.1 - 7, 8 1366
AK294410.1 - 4, 5 666
AK298534.1 - 4, 5 649
AL136853.1 - 7, 8 1382
AY364253.1 - 7, 8 1299
BC017464.1 - 7, 8 1329
BC028097.1 - 7, 8 1402 5242418
BF095127.1 - 2, 3 245

View Full Product Details