Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001011885.1 | NP_001011885.1 | 7 | 2718 | |
NM_025238.3 | NP_079514.1 | 8 | 2806 | ||
GenBank mRNA | AF257241.1 | - | 8 | 2685 | |
AF355402.1 | - | 8 | 2686 | ||
AL136853.1 | - | 8 | 2686 |