Gene Transcripts

Gene Symbol
MAPK14
Entrez Gene ID
1432
Gene Name
mitogen-activated protein kinase 14
Gene Aliases
CSBP, CSBP1, CSBP2, CSPB1, EXIP, Mxi2, PRKM14, PRKM15, RK, SAPK2A, p38, p38ALPHA
Chromosome Location
Chr. 6: 36027635 - 36122964 on Build GRCh38
UniGene ID
Hs.485233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001315.2 NP_001306.1 5, 6 939
NM_139012.2 NP_620581.1 5, 6 939
NM_139013.2 NP_620582.1 5, 6 939
NM_139014.2 NP_620583.1 5, 6 939
XM_006714998.3 XP_006715061.1 5, 6 414
XM_011514310.2 XP_011512612.2 5, 6 1207
XM_017010299.1 XP_016865788.1 5, 6 981
XM_017010300.1 XP_016865789.1 5, 6 981
XM_017010301.1 XP_016865790.1 5, 6 981
XM_017010302.1 XP_016865791.1 5, 6 981
XM_017010303.1 XP_016865792.1 5, 6 981
XM_017010304.1 XP_016865793.1 5, 6 981
GenBank mRNA AB074150.1 - 5, 6 458
AF100544.1 - 5, 6 473
AK291709.1 - 5, 6 834
AK303414.1 - 6, 7 503
BC000092.1 - 5, 6 796 3505234
BC031574.1 - 5, 6 795 5181064
BT006933.1 - 5, 6 458
CR536505.1 - 5, 6 458
FJ032367.1 - 5, 6 458
FJ032368.1 - 5, 6 458
L35263.1 - 5, 6 820
L35264.1 - 5, 6 666

View Full Product Details