Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007255.2 | NP_009186.1 | 6 | 1518 | |
XM_006714816.3 | XP_006714879.1 | 6 | 1450 | ||
XM_017008999.1 | XP_016864488.1 | 6 | 3641 | ||
GenBank mRNA | AB028600.1 | - | 6 | 1465 | |
AF142675.1 | - | 6 | 1458 | ||
AK022566.1 | - | 4 | 2525 | ||
AK023506.1 | - | 6 | 1461 | ||
AK095882.1 | - | 6 | 2122 | ||
AK123092.1 | - | 2 | 2244 | ||
BC007317.2 | - | 6 | 1518 | 3049998 | |
BC062983.1 | - | 6 | 1485 | 4577572 | |
BC072403.1 | - | 6 | 1507 | 6094074 |