Gene Transcripts
- Gene Symbol
- NFATC1
- Entrez Gene ID
- 4772
- Gene Name
- nuclear factor of activated T-cells 1
- Gene Aliases
- NF-ATC, NF-ATc1.2, NFAT2, NFATc
- Chromosome Location
- Chr. 18: 79395772 - 79529323 on Build GRCh38
- UniGene ID
- Hs.534074
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278669.1 |
NP_001265598.1
|
7, 8 |
2417 |
|
NM_001278670.1 |
NP_001265599.1
|
7, 8 |
2417 |
|
NM_001278672.1 |
NP_001265601.1
|
7, 8 |
2043 |
|
NM_001278673.1 |
NP_001265602.1
|
6, 7 |
944 |
|
NM_001278675.1 |
NP_001265604.1
|
7, 8 |
2043 |
|
NM_006162.4 |
NP_006153.2
|
7, 8 |
2417 |
|
NM_172387.2 |
NP_765975.1
|
7, 8 |
2043 |
|
NM_172388.2 |
NP_765976.1
|
6, 7 |
1318 |
|
NM_172389.2 |
NP_765977.1
|
7, 8 |
2043 |
|
NM_172390.2 |
NP_765978.1
|
7, 8 |
2417 |
|
GenBank mRNA |
AK292641.1 |
- |
7, 8 |
2259 |
|
AK293760.1 |
- |
6, 7 |
889 |
|
BC104753.1 |
- |
7, 8 |
2058 |
8143756 |
BC112243.1 |
- |
7, 8 |
2058 |
8327711 |
BC143457.1 |
- |
7, 8 |
1973 |
|
EU887559.1 |
- |
7, 8 |
1964 |
|
EU887560.1 |
- |
7, 8 |
1925 |
|
EU887561.1 |
- |
7, 8 |
1964 |
|
EU887562.1 |
- |
7, 8 |
1925 |
|
EU887563.1 |
- |
7, 8 |
1964 |
|
EU887564.1 |
- |
7, 8 |
1925 |
|
EU887565.1 |
- |
7, 8 |
1964 |
|
EU887566.1 |
- |
7, 8 |
1925 |
|
U08015.1 |
- |
7, 8 |
2203 |
|
U59736.1 |
- |
7, 8 |
2000 |
|
U80917.1 |
- |
7, 8 |
2333 |
|
U80918.1 |
- |
7, 8 |
1987 |
|
U80919.1 |
- |
6, 7 |
1146 |
|
View Full Product Details