Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001198696.1 | NP_001185625.1 | 7 | 867 | |
NM_032797.5 | NP_116186.1 | 7 | 864 | ||
GenBank mRNA | AF337957.1 | - | 6 | 668 | |
AF506757.1 | - | 7 | 864 | ||
AK027403.1 | - | 7 | 743 | ||
AK127353.1 | - | 7 | 2619 | ||
BC006121.1 | - | 6 | 672 | 3506309 | |
BC023601.2 | - | 7 | 851 | 4548458 |