Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000375.2 | NP_000366.1 | 8 | 765 | |
NM_001324036.1 | NP_001310965.1 | 8 | 765 | ||
NM_001324037.1 | NP_001310966.1 | 7 | 684 | ||
NM_001324038.1 | NP_001310967.1 | 7 | 684 | ||
NR_136675.1 | 6 | 584 | |||
NR_136676.1 | 8 | 765 | |||
NR_136677.1 | 8 | 765 | |||
NR_136678.1 | 5 | 495 | |||
XM_005270140.4 | XP_005270197.1 | 8 | 607 | ||
XM_011540127.1 | XP_011538429.1 | 8 | 607 | ||
XM_017016610.1 | XP_016872099.1 | 8 | 607 | ||
XM_017016611.1 | XP_016872100.1 | 8 | 563 | ||
XM_017016612.1 | XP_016872101.1 | 8 | 607 | ||
GenBank mRNA | AF230665.1 | - | 7 | 632 | |
AK314896.1 | - | 8 | 622 | ||
BC002573.2 | - | 8 | 724 | 3160463 | |
BI668388.1 | - | 5 | 435 | ||
J03824.1 | - | 8 | 695 | ||
KU178377.1 | - | 7 | 499 | ||
KU178378.1 | - | 6 | 418 |