Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256706.1 | NP_001243635.1 | 5 | 572 | |
NM_001256707.1 | NP_001243636.1 | 6 | 645 | ||
NM_001256708.1 | NP_001243637.1 | 5 | 778 | ||
NM_001256709.1 | NP_001243638.1 | 7 | 713 | ||
NM_001256710.1 | NP_001243639.1 | 6 | 610 | ||
NM_001256711.1 | NP_001243640.1 | 7 | 683 | ||
NM_001256712.1 | NP_001243641.1 | 4 | 440 | ||
NM_001318367.1 | NP_001305296.1 | 5 | 610 | ||
NM_014885.4 | NP_055700.2 | 5 | 561 | ||
NR_046345.1 | 3 | 324 | |||
XM_011531524.2 | XP_011529826.1 | 6 | 801 | ||
XM_011531531.2 | XP_011529833.1 | 4 | 467 | ||
XM_017007638.1 | XP_016863127.1 | 5 | 588 | ||
XM_017007639.1 | XP_016863128.1 | 6 | 1237 | ||
XM_017007640.1 | XP_016863129.1 | 5 | 629 | ||
XM_017007643.1 | XP_016863132.1 | 6 | 626 | ||
XM_017007644.1 | XP_016863133.1 | 6 | 600 | ||
GenBank mRNA | AB012109.1 | - | 5 | 559 | |
AF132794.1 | - | 5 | 516 | ||
AL080090.1 | - | 6 | 619 | ||
AV757106.2 | - | 4 | 499 | ||
BC005217.1 | - | 5 | 538 | 3929766 | |
BI488888.1 | - | 3 | 322 | ||
BI913723.1 | - | 5 | 611 | ||
BU175327.1 | - | 5 | 542 | ||
CN305215.1 | - | 5 | 478 | ||
CV023091.1 | - | 4 | 460 |