Gene Transcripts

Gene Symbol
B3GNT6
Entrez Gene ID
192134
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases
B3Gn-T6, BGnT-6, beta-1,3-Gn-T6, beta3Gn-T6
Chromosome Location
Chr. 11: 77034341 - 77041973 on Build GRCh38
UniGene ID
Hs.352622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138706.4 NP_619651.3 2 2205
GenBank mRNA AK127544.1 - 2 1788
AK172863.1 - 2 2130
BC025357.1 - 1 1746

View Full Product Details