Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001202554.1 | NP_001189483.1 | 4 | 1972 | |
NM_017837.3 | NP_060307.2 | 4 | 1646 | ||
GenBank mRNA | AK000484.1 | - | 4 | 1421 | |
AK056539.1 | - | 4 | 1965 | ||
AK097053.1 | - | 1 | 1071 | ||
AK225521.1 | - | 4 | 1421 | ||
AK301626.1 | - | 5 | 951 | ||
BC013568.1 | - | 4 | 1614 | 3890907 |