Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134734.1 | NP_001128206.1 | 3 | 1878 | |
NM_032884.4 | NP_116273.2 | 3 | 1009 | ||
GenBank mRNA | AK123355.1 | - | 3 | 1009 | |
BC007637.1 | - | 3 | 777 | 3529463 | |
BC064845.1 | - | 3 | 1160 | 30390219 |