Gene Transcripts

Gene Symbol
PATE1
Entrez Gene ID
160065
Gene Name
prostate and testis expressed 1
Gene Aliases
PATE
Chromosome Location
Chr. 11: 125746293 - 125749848 on Build GRCh38
UniGene ID
Hs.148565
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138294.2 NP_612151.1 5 647
GenBank mRNA AF462605.1 - 5 643
BC039863.1 - 5 645 5745088

View Full Product Details