Gene Transcripts

Gene Symbol
C11orf44
Entrez Gene ID
283171
Gene Name
chromosome 11 open reading frame 44
Gene Aliases
Chromosome Location
Chr. 11: 130672956 - 130717352 on Build GRCh38
UniGene ID
Hs.376151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271983.1 NP_001258912.1 3 697
GenBank mRNA AK096377.1 - 3 697

View Full Product Details