Gene Transcripts

Gene Symbol
C10orf53
Entrez Gene ID
282966
Gene Name
chromosome 10 open reading frame 53
Gene Aliases
Chromosome Location
Chr. 10: 49679638 - 49708910 on Build GRCh38
UniGene ID
Hs.131287
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182554.2 NP_872360.2 3 331
GenBank mRNA BC028127.1 - 3 283

View Full Product Details