Gene Transcripts
- Gene Symbol
- AXDND1
- Entrez Gene ID
- 126859
- Gene Name
- axonemal dynein light chain domain containing 1
- Gene Aliases
- C1orf125
- Chromosome Location
- Chr. 1: 179365705 - 179554735 on Build GRCh38
- UniGene ID
- Hs.658505
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144696.5 |
NP_653297.3
|
8, 9 |
1132 |
|
NR_073544.1 |
|
8, 9 |
1006 |
|
XM_011509165.1 |
XP_011507467.1
|
8, 9 |
1111 |
|
XM_011509166.2 |
XP_011507468.1
|
8, 9 |
1158 |
|
XM_011509167.2 |
XP_011507469.1
|
8, 9 |
1273 |
|
XM_011509168.1 |
XP_011507470.1
|
8, 9 |
1115 |
|
XM_011509169.1 |
XP_011507471.1
|
7, 8 |
904 |
|
XM_011509170.1 |
XP_011507472.1
|
8, 9 |
1111 |
|
XM_011509171.1 |
XP_011507473.1
|
7, 8 |
929 |
|
XM_011509174.1 |
XP_011507476.1
|
8, 9 |
1111 |
|
XM_011509175.1 |
XP_011507477.1
|
8, 9 |
1111 |
|
XM_011509176.1 |
XP_011507478.1
|
8, 9 |
1111 |
|
XM_011509178.2 |
XP_011507480.1
|
8, 9 |
1111 |
|
XM_011509179.2 |
XP_011507481.1
|
3, 4 |
219 |
|
XM_011509180.1 |
XP_011507482.1
|
8, 9 |
1111 |
|
GenBank mRNA |
AK057502.1 |
- |
6, 7 |
499 |
|
AK302972.1 |
- |
8, 9 |
1111 |
|
AL833914.1 |
- |
6, 7 |
559 |
|
BC146823.1 |
- |
8, 9 |
1007 |
8860438 |
BX647935.1 |
- |
8, 9 |
1133 |
|
View Full Product Details