Gene Transcripts
- Gene Symbol
- AFAP1
- Entrez Gene ID
- 60312
- Gene Name
- actin filament associated protein 1
- Gene Aliases
- AFAP, AFAP-110, AFAP110
- Chromosome Location
- Chr. 4: 7758713 - 7939926 on Build GRCh38
- UniGene ID
- Hs.529369
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134647.1 |
NP_001128119.1
|
2, 3 |
408 |
|
NM_198595.2 |
NP_940997.1
|
2, 3 |
408 |
|
XM_006713909.3 |
XP_006713972.1
|
2, 3 |
291 |
|
XM_011513544.2 |
XP_011511846.1
|
2, 3 |
461 |
|
XM_017008535.1 |
XP_016864024.1
|
1, 2 |
137 |
|
XM_017008536.1 |
XP_016864025.1
|
1, 2 |
137 |
|
GenBank mRNA |
AK290807.1 |
- |
2, 3 |
235 |
|
AK297631.1 |
- |
2, 3 |
201 |
|
AL706541.1 |
- |
1, 2 |
353 |
|
BC032777.1 |
- |
1, 2 |
159 |
4914388 |
BI561136.1 |
- |
2, 3 |
408 |
|
DA605903.1 |
- |
2, 3 |
341 |
|
DB450895.1 |
- |
2, 3 |
408 |
|
View Full Product Details