Gene Transcripts
- Gene Symbol
- ALS2CR11
- Entrez Gene ID
- 151254
- Gene Name
- amyotrophic lateral sclerosis 2 chromosome region candidate 11
- Gene Aliases
- Chromosome Location
- Chr. 2: 201487421 - 201619182 on Build GRCh38
- UniGene ID
- Hs.335788
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK058080.1 |
- |
11 |
1147 |
|
BC030659.2 |
- |
11 |
1168 |
4838801 |
View Full Product Details