Gene Transcripts

Gene Symbol
LRRC34
Entrez Gene ID
151827
Gene Name
leucine rich repeat containing 34
Gene Aliases
Chromosome Location
Chr. 3: 169793006 - 169812904 on Build GRCh38
UniGene ID
Hs.591289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172779.1 NP_001166250.1 11 1590
NM_153353.4 NP_699184.2 10 1494
XM_006713508.3 XP_006713571.1 11 1320
XM_011512442.2 XP_011510744.1 11 1371
XM_017005746.1 XP_016861235.1 11 1545
GenBank mRNA AK130856.1 - 4 563
AK293661.1 - 11 1468
AK302466.1 - 11 1352
BC032841.2 - 10 1233 4826965

View Full Product Details