Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317002.1 | NP_001303931.1 | 8 | 1094 | |
NM_001317003.1 | NP_001303932.1 | 6 | 1204 | ||
NM_001317004.1 | NP_001303933.1 | 7 | 1005 | ||
NM_001317005.1 | NP_001303934.1 | 7 | 1265 | ||
NM_001317006.1 | NP_001303935.1 | 5 | 1133 | ||
NM_177964.4 | NP_808879.2 | 7 | 1276 | ||
XM_005246306.4 | XP_005246363.1 | 9 | 1246 | ||
XM_006712279.2 | XP_006712342.1 | 7 | 1098 | ||
XM_006712280.3 | XP_006712343.1 | 9 | 1244 | ||
XM_006712281.3 | XP_006712344.1 | 7 | 1018 | ||
XM_011510620.2 | XP_011508922.1 | 9 | 1119 | ||
XM_011510623.2 | XP_011508925.1 | 8 | 1092 | ||
XM_011510625.1 | XP_011508927.1 | 10 | 1323 | ||
XM_017003364.1 | XP_016858853.1 | 8 | 1104 | ||
XM_017003365.1 | XP_016858854.1 | 7 | 1031 | ||
XM_017003366.1 | XP_016858855.1 | 7 | 1075 | ||
GenBank mRNA | AF435957.1 | - | 7 | 1005 | |
BC018203.1 | - | 6 | 952 | 3861272 | |
BC040176.2 | - | 7 | 1276 | 4794338 |