Gene Transcripts
- Gene Symbol
- WIPI2
- Entrez Gene ID
- 26100
- Gene Name
- WD repeat domain, phosphoinositide interacting 2
- Gene Aliases
- ATG18B, Atg21, CGI-50, WIPI-2
- Chromosome Location
- Chr. 7: 5190204 - 5233855 on Build GRCh38
- UniGene ID
- Hs.122363
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033518.1 |
NP_001028690.1
|
8, 9 |
954 |
|
NM_001033519.1 |
NP_001028691.1
|
7, 8 |
900 |
|
NM_001033520.1 |
NP_001028692.1
|
5, 6 |
793 |
|
NM_001278299.1 |
NP_001265228.1
|
8, 9 |
1128 |
|
NM_015610.3 |
NP_056425.1
|
8, 9 |
954 |
|
NM_016003.3 |
NP_057087.2
|
7, 8 |
900 |
|
XM_006715685.1 |
XP_006715748.1
|
6, 7 |
786 |
|
XM_017011952.1 |
XP_016867441.1
|
6, 7 |
786 |
|
GenBank mRNA |
AF151808.1 |
- |
5, 6 |
792 |
|
AK023041.1 |
- |
8, 9 |
914 |
|
AK024279.1 |
- |
7, 8 |
879 |
|
AK315468.1 |
- |
8, 9 |
1074 |
|
AL080155.1 |
- |
8, 9 |
955 |
|
AY691425.1 |
- |
7, 8 |
684 |
|
AY691426.1 |
- |
5, 6 |
724 |
|
BC004116.2 |
- |
8, 9 |
897 |
3842487 |
BC007596.1 |
- |
7, 8 |
867 |
3346710 |
BC021012.1 |
- |
7, 8 |
819 |
|
BC021068.1 |
- |
7, 8 |
819 |
|
BC021200.2 |
- |
8, 9 |
895 |
3957981 |
CR533530.1 |
- |
8, 9 |
738 |
|
View Full Product Details