Gene Transcripts

Gene Symbol
ARSJ
Entrez Gene ID
79642
Gene Name
arylsulfatase family member J
Gene Aliases
Chromosome Location
Chr. 4: 113899929 - 113980125 on Build GRCh38
UniGene ID
Hs.22895
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024590.3 NP_078866.3 2 1391
XM_005263212.4 XP_005263269.1 2 1349
XM_011532238.2 XP_011530540.1 3 728
XM_011532239.2 XP_011530541.1 4 266
XM_017008592.1 XP_016864081.1 3 1431
XM_017008593.1 XP_016864082.1 6 1986
XM_017008594.1 XP_016864083.1 5 1553
XM_017008595.1 XP_016864084.1 6 1666
XM_017008596.1 XP_016864085.1 6 1646
XM_017008597.1 XP_016864086.1 6 1677
XM_017008598.1 XP_016864087.1 4 1503
XM_017008599.1 XP_016864088.1 4 1512
XM_017008600.1 XP_016864089.1 3 216
GenBank mRNA AK307532.1 - 4 1033
AM049401.1 - 2 665
AY358647.1 - 2 1393
AY875938.1 - 2 503
BC033156.1 - 2 295
BC089445.1 - 2 761
BC132879.1 - 2 591 40146901
BC132881.1 - 2 591 40146903
BC144265.1 - 2 591 9052785
BC151213.1 - 3 1607

View Full Product Details