Gene Transcripts

Gene Symbol
NUP43
Entrez Gene ID
348995
Gene Name
nucleoporin 43
Gene Aliases
bA350J20.1, p42
Chromosome Location
Chr. 6: 149724315 - 149746572 on Build GRCh38
UniGene ID
Hs.510375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198887.2 NP_942590.1 2 272
NR_104456.1 2 272
XM_005266960.4 XP_005267017.1 2 222
XM_005266961.3 XP_005267018.1 2 222
XM_005266962.3 XP_005267019.1 2 222
XM_011535798.2 XP_011534100.1 2 222
XM_011535799.2 XP_011534101.1 2 222
GenBank mRNA AF514997.1 - 2 205
AK074311.1 - 228
AK095994.1 - 2 228
AK301027.1 - 2 222
AK304244.1 - 2 204
AK304505.1 - 2 260
BC065028.1 - 2 198 6042215
BX647097.1 - 2 222
CB989591.1 - 2 269
DC324759.1 - 2 272

View Full Product Details