Gene Transcripts

Gene Symbol
MTURN
Entrez Gene ID
222166
Gene Name
maturin, neural progenitor differentiation regulator homolog (Xenopus)
Gene Aliases
C7orf41, Ells1
Chromosome Location
Chr. 7: 30134936 - 30162765 on Build GRCh38
UniGene ID
Hs.200100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152793.2 NP_690006.2 3 975
GenBank mRNA AL834391.1 - 3 909
AY054121.1 - 3 771
BC062423.1 - 3 726

View Full Product Details