Gene Transcripts
- Gene Symbol
- C7orf33
- Entrez Gene ID
- 202865
- Gene Name
- chromosome 7 open reading frame 33
- Gene Aliases
- Chromosome Location
- Chr. 7: 148590565 - 148615860 on Build GRCh38
- UniGene ID
- Hs.352212
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145304.3 |
NP_660347.1
|
2, 3 |
828 |
|
GenBank mRNA |
BC021251.2 |
- |
2, 3 |
828 |
|
View Full Product Details